siRNA Details
Gene Symbol: TCEB3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038280.1 | 4 | 828 | |||
GenBank mRNA | AL049675.1 | - | 4 | 818 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124839.1 | - | 1 | 2206 | Chr. 1 - 23760519 - 23762843 [-] on Build GRCh38 |
