siRNA Details
Gene Symbol: LINC01563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110895.1 | 3 | 743 | |||
NR_110896.1 | 2 | 619 | ||||
GenBank mRNA | AB593144.1 | - | 3 | 708 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CV569314.1 | - | 2 | 479 | Chr. 17 - 21075696 - 21090566 [+] on Build GRCh38 |
