siRNA Details
Gene Symbol: LINC01563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110895.1 | 1 | 156 | |||
NR_110896.1 | 1 | 156 | ||||
GenBank mRNA | AB593144.1 | - | 1 | 122 | ||
AV723259.1 | - | 1 | 156 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110895.1 | 1 | 156 | |||
NR_110896.1 | 1 | 156 | ||||
GenBank mRNA | AB593144.1 | - | 1 | 122 | ||
AV723259.1 | - | 1 | 156 |