siRNA Details
Gene Symbol: CBR3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038892.1 | 2 | 111 | |||
NR_038893.1 | 2 | 111 | ||||
NR_038894.1 | 2 | 111 | ||||
GenBank mRNA | AB004851.1 | - | 2 | 103 | ||
AL542398.3 | - | 2 | 98 | |||
BC037580.1 | - | 2 | 110 | |||
BC038671.1 | - | 2 | 4032 | |||
BC047014.1 | - | 2 | 110 | |||
BI836686.1 | - | 2 | 111 |
