siRNA Details
Gene Symbol: LINC00421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034026.1 | 2 | 971 | |||
GenBank mRNA | BC022083.2 | - | 2 | 971 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034026.1 | 2 | 971 | |||
GenBank mRNA | BC022083.2 | - | 2 | 971 |