siRNA Details
Gene Symbol: LOC100996263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121626.1 | 2 | 240 | |||
GenBank mRNA | BU568889.1 | - | 2 | 332 | ||
BX088863.1 | - | 2 | 181 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121626.1 | 2 | 240 | |||
GenBank mRNA | BU568889.1 | - | 2 | 332 | ||
BX088863.1 | - | 2 | 181 |