siRNA Details
Gene Symbol: ZEB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024284.1 | 2 | 264 | |||
GenBank mRNA | AK026657.1 | - | 1 | 224 | ||
AK296146.1 | - | 1 | 517 | |||
BC042008.1 | - | 1 | 554 | |||
BX446976.2 | - | 2 | 185 | |||
DB121883.1 | - | 2 | 265 | |||
DB127940.1 | - | 2 | 226 |
