siRNA Details
Gene Symbol: DLEU7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046551.1 | 1 | 116 | |||
GenBank mRNA | BC035769.1 | - | 1 | 126 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM454738.1 | - | 1 | 115 | Chr. 13 - 50807856 - 50849031 [+] on Build GRCh38 |
