siRNA Details
Gene Symbol: LOC101928973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125966.1 | 3 | 400 | |||
GenBank mRNA | BC039493.1 | - | 3 | 400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM551928.1 | - | 3 | 399 | Chr. 1 - 181174485 - 181182208 [+] on Build GRCh38 |
