siRNA Details
Gene Symbol: TTN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038272.1 | 10 | 2530 | |||
GenBank mRNA | BC041357.1 | - | 10 | 2538 | ||
BC070113.1 | - | 10 | 2533 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TTN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038272.1 | 10 | 2530 | |||
GenBank mRNA | BC041357.1 | - | 10 | 2538 | ||
BC070113.1 | - | 10 | 2533 |