siRNA Details
Gene Symbol: TTN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038271.1 | 1 | 338 | |||
NR_038272.1 | 2 | 111 | ||||
GenBank mRNA | AK093733.1 | - | 1 | 305 | ||
BC041357.1 | - | 2 | 119 | |||
BC070113.1 | - | 2 | 114 | |||
DB229458.1 | - | 1 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB108507.1 | - | 1 | 305 | Chr. 2 - 178523224 - 178597615 [+] on Build GRCh38 |
