siRNA Details
Gene Symbol: GS1-124K5.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134570.1 | 2 | 577 | |||
NR_134571.1 | 2 | 390 | ||||
NR_134572.1 | 2 | 386 | ||||
NR_134573.1 | 2 | 161 | ||||
GenBank mRNA | CN310937.1 | - | 1 | 59 | ||
CT005114.1 | - | 2 | 375 | |||
HY009490.1 | - | 2 | 388 | |||
HY053797.1 | - | 2 | 162 |
