siRNA Details
Gene Symbol: LINC01502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034016.1 | 2 | 278 | |||
NR_034017.1 | 2 | 278 | ||||
NR_109814.1 | 2 | 278 | ||||
GenBank mRNA | AK098241.1 | - | 2 | 252 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034016.1 | 2 | 278 | |||
NR_034017.1 | 2 | 278 | ||||
NR_109814.1 | 2 | 278 | ||||
GenBank mRNA | AK098241.1 | - | 2 | 252 |