siRNA Details
Gene Symbol: MGC27382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027310.2 | 1, 2 | 139 | |||
GenBank mRNA | AK091757.1 | - | 1, 2 | 139 | ||
BC015860.1 | - | 1, 2 | 120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG536943.1 | - | 1, 2 | 117 | Chr. 1 - 78229622 - 78342620 [+] on Build GRCh38 |
