siRNA Details
Gene Symbol: LOC101928661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125860.1 | 2, 3 | 209 | |||
GenBank mRNA | BC034912.1 | - | 2, 3 | 208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI465288.1 | - | 2, 3 | 208 | Chr. 6 - 146595951 - 146598931 [-] on Build GRCh38 |
