siRNA Details
Gene Symbol: UNC5B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038453.1 | 1 | 166 | |||
GenBank mRNA | CK819868.1 | - | 1 | 166 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM142058.1 | - | 1 | 150 | Chr. 10 - 71217374 - 71218212 [-] on Build GRCh38 |
