siRNA Details
Gene Symbol: FRY-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103839.1 | 4 | 768 | |||
GenBank mRNA | AF086419.1 | - | 1 | 201 | ||
BC035084.1 | - | 5 | 773 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FRY-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103839.1 | 4 | 768 | |||
GenBank mRNA | AF086419.1 | - | 1 | 201 | ||
BC035084.1 | - | 5 | 773 |