siRNA Details
Gene Symbol: TEX36-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023362.1 | 5 | 720 | |||
GenBank mRNA | BC033977.2 | - | 5 | 720 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI559430.1 | - | 5 | 725 | Chr. 10 - 125574371 - 125578445 [+] on Build GRCh38 |
