siRNA Details
Gene Symbol: LINC00649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038883.2 | 1 | 564 | |||
NR_134559.1 | 1 | 564 | ||||
GenBank mRNA | AK311545.1 | - | 1 | 564 | ||
AL532366.3 | - | 1 | 49 | |||
BX102617.1 | - | 1 | 523 | |||
DA374118.1 | - | 1 | 474 |
