siRNA Details
Gene Symbol: GBAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131972.1 | 2 | 221 | |||
GenBank mRNA | AA281900.1 | - | 2 | 194 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GBAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131972.1 | 2 | 221 | |||
GenBank mRNA | AA281900.1 | - | 2 | 194 |