siRNA Details
Gene Symbol: GBAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131972.1 | 1 | 101 | |||
GenBank mRNA | AA281900.1 | - | 1 | 74 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG531335.1 | - | 1 | 86 | Chr. 1 - 151347024 - 151348027 [+] on Build GRCh38 |
