siRNA Details
Gene Symbol: LINC01053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120427.1 | 3 | 589 | |||
GenBank mRNA | AB593160.1 | - | 2 | 3173 | ||
BU176203.1 | - | 3 | 566 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120427.1 | 3 | 589 | |||
GenBank mRNA | AB593160.1 | - | 2 | 3173 | ||
BU176203.1 | - | 3 | 566 |