siRNA Details
Gene Symbol: CD81-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC019904.1 | - | 2 | 432 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CD81-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC019904.1 | - | 2 | 432 |