siRNA Details
Gene Symbol: PAXBP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038879.1 | 1 | 1488 | |||
NR_038880.1 | 1 | 1488 | ||||
GenBank mRNA | AK056384.1 | - | 1 | 1487 | ||
BC038361.1 | - | 1 | 1463 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PAXBP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038879.1 | 1 | 1488 | |||
NR_038880.1 | 1 | 1488 | ||||
GenBank mRNA | AK056384.1 | - | 1 | 1487 | ||
BC038361.1 | - | 1 | 1463 |