siRNA Details
Gene Symbol: ST6GAL2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX112866.1 | - | 2 | 266 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ST6GAL2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX112866.1 | - | 2 | 266 |