siRNA Details
Gene Symbol: MYLK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038266.2 | 3 | 790 | |||
NR_121654.1 | 4 | 697 | ||||
GenBank mRNA | AV744989.1 | - | 2 | 564 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MYLK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038266.2 | 3 | 790 | |||
NR_121654.1 | 4 | 697 | ||||
GenBank mRNA | AV744989.1 | - | 2 | 564 |