siRNA Details
Gene Symbol: C21orf91-OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038870.1 | 3 | 254 | |||
NR_038871.1 | 4 | 319 | ||||
GenBank mRNA | AY063452.1 | - | 4 | 319 | ||
AY063453.1 | - | 3 | 254 | |||
BX099338.1 | - | 3 | 255 | |||
CR745563.1 | - | 3 | 253 |
