siRNA Details
Gene Symbol: PTPRG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038281.1 | 5 | 1955 | |||
NR_038282.1 | 5 | 1941 | ||||
NR_038283.1 | 4 | 1859 | ||||
GenBank mRNA | AK022901.1 | - | 3 | 1711 | ||
AK094634.1 | - | 4 | 1835 | |||
BC041886.1 | - | 5 | 1885 |
