siRNA Details
Gene Symbol: PTPRG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038281.1 | 2 | 118 | |||
NR_038282.1 | 2 | 121 | ||||
NR_038283.1 | 2 | 118 | ||||
GenBank mRNA | AK094634.1 | - | 2 | 94 | ||
AK307209.1 | - | 2 | 106 | |||
AK308660.1 | - | 2 | 118 | |||
BC041886.1 | - | 2 | 65 |
