siRNA Details
Gene Symbol: LOC105374177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135546.1 | 3 | 1385 | |||
GenBank mRNA | BC038760.1 | - | 3 | 1383 | ||
DB507682.1 | - | 3 | 334 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105374177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135546.1 | 3 | 1385 | |||
GenBank mRNA | BC038760.1 | - | 3 | 1383 | ||
DB507682.1 | - | 3 | 334 |