siRNA Details
Gene Symbol: ADAMTS9-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038264.1 | 2 | 488 | |||
GenBank mRNA | AK057923.1 | - | 2 | 256 | ||
AL832788.1 | - | 2 | 449 | |||
BC040632.1 | - | 2 | 230 | |||
DA927615.1 | - | 2 | 490 |
