siRNA Details
Gene Symbol: PRR34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027034.1 | 2 | 105 | |||
GenBank mRNA | CR986432.1 | - | 2 | 190 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRR34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027034.1 | 2 | 105 | |||
GenBank mRNA | CR986432.1 | - | 2 | 190 |