siRNA Details
Gene Symbol: KLHL6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046687.1 | 2 | 459 | |||
GenBank mRNA | BX091913.1 | - | 2 | 459 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KLHL6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046687.1 | 2 | 459 | |||
GenBank mRNA | BX091913.1 | - | 2 | 459 |