siRNA Details
Gene Symbol: SOX2-OT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_075091.1 | 4 | 350 | |||
NR_075092.1 | 4 | 350 | ||||
NR_075093.1 | 3 | 326 | ||||
GenBank mRNA | BC041898.2 | - | 1 | 131 | ||
DA281835.1 | - | 4 | 350 | |||
DA310380.1 | - | 3 | 326 | |||
JQ408703.1 | - | 4 | 349 | |||
JX110620.1 | - | 3 | 441 | |||
JX504683.1 | - | 4 | 403 | |||
KJ415055.1 | - | 1 | 30 |
