siRNA Details
Gene Symbol: LOC101926934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134642.1 | 1, 2 | 242 | |||
GenBank mRNA | BX114358.1 | - | 1, 2 | 243 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101926934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134642.1 | 1, 2 | 242 | |||
GenBank mRNA | BX114358.1 | - | 1, 2 | 243 |