siRNA Details
Gene Symbol: LOC101927668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110114.1 | 3, 4 | 207 | |||
GenBank mRNA | AI792720.1 | - | 3, 4 | 155 | ||
CK904648.1 | - | 3, 4 | 207 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110114.1 | 3, 4 | 207 | |||
GenBank mRNA | AI792720.1 | - | 3, 4 | 155 | ||
CK904648.1 | - | 3, 4 | 207 |