siRNA Details
Gene Symbol: LOC101927668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110114.1 | 4 | 332 | |||
GenBank mRNA | AI792720.1 | - | 4 | 280 | ||
CK904648.1 | - | 4 | 332 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110114.1 | 4 | 332 | |||
GenBank mRNA | AI792720.1 | - | 4 | 280 | ||
CK904648.1 | - | 4 | 332 |