siRNA Details
Gene Symbol: TPTE2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002815.2 | 8, 9 | 2430 | |||
GenBank mRNA | AL137411.1 | - | 12, 13 | 1424 | ||
BC045761.1 | - | 8, 9 | 2429 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TPTE2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002815.2 | 8, 9 | 2430 | |||
GenBank mRNA | AL137411.1 | - | 12, 13 | 1424 | ||
BC045761.1 | - | 8, 9 | 2429 |