siRNA Details
Gene Symbol: ANKRD30BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043208.2 | - | 5 | 567 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANKRD30BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043208.2 | - | 5 | 567 |