siRNA Details
Gene Symbol: MNX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038835.1 | 1 | 419 | |||
GenBank mRNA | BC022831.1 | - | 1 | 370 | ||
BC033117.1 | - | 1 | 370 | |||
BU506764.1 | - | 1 | 422 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MNX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038835.1 | 1 | 419 | |||
GenBank mRNA | BC022831.1 | - | 1 | 370 | ||
BC033117.1 | - | 1 | 370 | |||
BU506764.1 | - | 1 | 422 |