siRNA Details
Gene Symbol: LOC101929717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110823.1 | 4 | 2014 | |||
GenBank mRNA | AK096271.1 | - | 4 | 2106 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110823.1 | 4 | 2014 | |||
GenBank mRNA | AK096271.1 | - | 4 | 2106 |