siRNA Details
Gene Symbol: LOC101929717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110823.1 | 3 | 291 | |||
GenBank mRNA | AK096271.1 | - | 3 | 386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA697791.1 | - | 3 | 386 | Chr. 3 - 113144284 - 113167819 [-] on Build GRCh38 |
