siRNA Details
Gene Symbol: WDR86-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034012.2 | 5 | 704 | |||
NR_034013.2 | 4 | 619 | ||||
GenBank mRNA | AK095986.1 | - | 4 | 544 | ||
BC043579.1 | - | 4 | 495 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WDR86-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034012.2 | 5 | 704 | |||
NR_034013.2 | 4 | 619 | ||||
GenBank mRNA | AK095986.1 | - | 4 | 544 | ||
BC043579.1 | - | 4 | 495 |