siRNA Details
Gene Symbol: LOC101927123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110147.1 | 4 | 353 | |||
GenBank mRNA | BC015846.1 | - | 4 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG483991.1 | - | 4 | 354 | Chr. 3 - 127322526 - 127390670 [-] on Build GRCh38 |
