siRNA Details
Gene Symbol: PEX5L-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110059.1 | 3 | 361 | |||
NR_110060.1 | 3 | 244 | ||||
NR_110061.1 | 4 | 298 | ||||
GenBank mRNA | BG186146.1 | - | 1 | 46 | ||
HY015924.1 | - | 4 | 291 | |||
HY048179.1 | - | 3 | 362 | |||
HY052832.1 | - | 3 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX104086.1 | - | 3 | 326 | Chr. 3 - 179898229 - 179921745 [+] on Build GRCh38 |
