siRNA Details
Gene Symbol: PEX5L-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110059.1 | 3 | 613 | |||
NR_110060.1 | 3 | 496 | ||||
NR_110061.1 | 4 | 550 | ||||
GenBank mRNA | BG186146.1 | - | 1 | 298 | ||
HY015924.1 | - | 4 | 543 |
