siRNA Details
Gene Symbol: LINC00698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027104.1 | 7 | 1483 | |||
GenBank mRNA | BC039502.1 | - | 6 | 1075 | ||
BC043407.1 | - | 9 | 1604 | |||
BC104474.1 | - | 1 | 385 | |||
BC104475.1 | - | 1 | 385 |
