siRNA Details
Gene Symbol: GABPB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024490.1 | 1 | 2126 | |||
GenBank mRNA | BX537481.1 | - | 2 | 279 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GABPB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024490.1 | 1 | 2126 | |||
GenBank mRNA | BX537481.1 | - | 2 | 279 |