siRNA Details
Gene Symbol: ALKBH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038907.1 | 6 | 1453 | |||
GenBank mRNA | AK097541.1 | - | 6 | 1436 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ALKBH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038907.1 | 6 | 1453 | |||
GenBank mRNA | AK097541.1 | - | 6 | 1436 |