siRNA Details
Gene Symbol: LOC101928445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110560.1 | 2 | 171 | |||
GenBank mRNA | AK128666.1 | - | 2 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB258449.1 | - | 2 | 171 | Chr. 5 - 179602247 - 179603741 [-] on Build GRCh38 |
